Learn More
Aat BioquestSupplier Diversity Partner ReadiLink™ Protein Conjugation Stop Buffer  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 5400
ReadiLink™ Protein Conjugation Stop Buffer is used to stop the protein conjugation reactions that use succinimidyl ester, sulfonyl chloride or isothiocyanate. It can be used directly without dilution.
Catalog No. 50-195-3284