Learn More
Connecticut Clean Room™ PolyTack Roll Mop and Accessories
Perfect for cleaning walls, floors, ceiling and hard to reach areas
Manufacturer:  Connecticut Clean Room™ 4114R
 View more versions of this product
Catalog No. 19-047-519
Add to cart
Description
Description
  • Helps keep areas free of contamination with easy to use hand or pole attachments
  • Special anti-microbial agents provide long-lasting protection against bacterial growth
  • Plastic inner core insures cleanliness
  • Refills and replacement handles, mops and tacky rollers available
  • 19-823-738 is a film roll only; it does not include  a handle or extension pole
Specifications
Specifications
Safety
N/A
For removing particulate matter from the cleanroom
Yes
Mop Refill
35.5cm (14 in.)