Learn More
Cole-Parmer Barbed Fittings; Reducing Connector, Stainless Steel, 1/2" x 1/4" Tubing ID  DFS Item
Manufacturer:  Cole-Parmer 3062011
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. 316 stainless steel material features very good chemical resistance with a wide temperature range. Sterilize via ethylene oxide, autoclave, or gamma irradiation.
Catalog No. 50-629-551