Learn More
Cole-Parmer Masterflex Fitting, 316 Stainless Steel, Straight, Hose Barb Union, 3/8" ID  DFS Item
Manufacturer:  Cole-Parmer 3062005
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-260