Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 32/GJB1 Rat anti-Mouse, Rat, DyLight 405, Clone: R5.21C, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275945V
Description
Connexin 32/GJB1 Monoclonal antibody specifically detects Connexin 32/GJB1 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).Specifications
Connexin 32/GJB1 | |
R5.21C | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Conjugated, Purified | |
GJB1 | |
Mouse liver DOC-JR-plasma membranes | |
Protein G purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Mouse, Rat |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
DyLight 405 | |
50 mM Sodium Borate with 0.05% Sodium Azide | |
CMTX, Connexin-32, Cx32, CX32beta 1, 32kD (connexin 32, Charcot-Marie-Toothneuropathy, X-linked), gap junction beta-1 protein, gap junction protein, beta 1, 32kDa, gap junction protein, beta 1, 32kDa (connexin 32), gap junction protein, beta 1, 32kDa (connexin 32, Charcot-Marie-Toothneuropathy, X-linked) | |
Rat | |
IgG2a | |
100 μL | |
Primary | |
2705.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title