Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 32/GJB1 Rat anti-Mouse, Rat, DyLight™ 755, Clone: R5.21C, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275945IR

Catalog No. NBP275945IR


Add to cart

Description

Description

Connexin 32/GJB1 Monoclonal antibody specifically detects Connexin 32/GJB1 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
Specifications

Specifications

Connexin 32/GJB1
R5.21C
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Conjugated, Purified
GJB1
Mouse liver DOC-JR-plasma membranes
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight 755
50 mM Sodium Borate with 0.05% Sodium Azide
CMTX, Connexin-32, Cx32, CX32beta 1, 32kD (connexin 32, Charcot-Marie-Toothneuropathy, X-linked), gap junction beta-1 protein, gap junction protein, beta 1, 32kDa, gap junction protein, beta 1, 32kDa (connexin 32), gap junction protein, beta 1, 32kDa (connexin 32, Charcot-Marie-Toothneuropathy, X-linked)
Rat
IgG2a
100 μL
Primary
2705.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit