Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 36/GJD2 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31039525UL

 View more versions of this product

Catalog No. NB125692


Description

Description

Connexin 36/GJD2 Polyclonal specifically detects Connexin 36/GJD2 in Rat samples. It is validated for Western Blot.
Specifications

Specifications

Connexin 36/GJD2
Unconjugated
PBS buffer, 2% sucrose
Connexin-36, Cx36, CX36connexin 36, Gap junction alpha-9 protein, gap junction delta-2 protein, gap junction protein, alpha 9, 36kDa, gap junction protein, delta 2, 36kDa, GJA9connexin-36, MGC138315, MGC138319
The immunogen is a synthetic peptide directed towards the middle region of Rat Connexin 36/GJD2 (NP_062154). Peptide sequence GGGGSGGSKREDKKLQNAIVNGVLQNTETTSKETEPDCLEVKELAPHPSG
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Rat
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
57369
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit