Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 36/GJD2 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen Connexin 36/GJD2
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125692
View Documents
Novus Biologicals
NBP31039525UL
25 μg
Each for $106.19
Add to cart
 
NB125691
View Documents
Novus Biologicals
NBP310395100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

Connexin 36/GJD2 Polyclonal antibody specifically detects Connexin 36/GJD2 in Rat samples. It is validated for Western Blot
Specifications

Specifications

Connexin 36/GJD2
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
57369
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Rat
Connexin-36, Cx36, CX36connexin 36, Gap junction alpha-9 protein, gap junction delta-2 protein, gap junction protein, alpha 9, 36kDa, gap junction protein, delta 2, 36kDa, GJA9connexin-36, MGC138315, MGC138319
The immunogen is a synthetic peptide directed towards the middle region of Rat Connexin 36/GJD2 (NP_062154). Peptide sequence GGGGSGGSKREDKKLQNAIVNGVLQNTETTSKETEPDCLEVKELAPHPSG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit