Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 40/GJA5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Connexin 40/GJA5
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095541
View Documents
Novus Biologicals
NBP303590100UL
100 μg
Each for $320.71
Add to cart
 
NB095542
View Documents
Novus Biologicals
NBP30359020UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Connexin 40/GJA5 Polyclonal antibody specifically detects Connexin 40/GJA5 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

Connexin 40/GJA5
Unconjugated
Rabbit
Human
alpha 5, 40kD (connexin 40), connexin-40, Cx40, gap junction alpha-5 protein, gap junction protein, alpha 5, 40kDa, gap junction protein, alpha 5, 40kDa (connexin 40), MGC11185
Recombinant fusion protein containing a sequence corresponding to amino acids 229-358 of human Connexin 40/GJA5 (NP_859054.1).
Polyclonal
Affinity purified
Immunofluorescence
Purified
RUO
PBS with 50% glycerol, pH7.3.
2702
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit