Learn More
Connexin 43/GJA1 Mouse anti-Human, Alexa Fluor 594, Clone: 578618, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB7737T100UG
Catalog No. FAB7737T100
Add to cart
This item is not returnable. View return policy
Description
Description
Connexin 43/GJA1 Monoclonal antibody specifically detects Connexin 43/GJA1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Connexin 43/GJA1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P17302
GJA1
Mouse myeloma cell line NS0 transfected with human Connexin 43/GJA1 Accession # P17302
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Connexin 43/GJA1. Stains human Connexin 43/GJA1 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
578618
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJA1, GJAL, HSS, ODD, ODDD, ODOD, SDTY3
Mouse
IgG2a
RUO
Primary
2697
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)