Learn More
Connexin-43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153861AP
Catalog No. 50-173-0192
Add to cart
Description
Description
This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. The encoded protein is the major protein of gap junctions in the heart that are thought to have a crucial role in the synchronized contraction of the heart and in embryonic development. A related intronless pseudogene has been mapped to chromosome 5. Mutations in this gene have been associated with oculodentodigital dysplasia and heart malformations.
Specifications
Specifications
Connexin-43
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08050, P17302, P23242
Connexin-43, CX43, DFNB38, Gap junction alpha 1 protein, GJA1, GJAL, ODDD
Rabbit
IgG
150 μL
-20°C
Primary
14609, 24392, 2697
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
GJA1
Liquid
Gja1
Connexin-43 Fusion Protein Ag6698
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat