Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 46/GJA3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31086925UL

 View more versions of this product

Catalog No. NB126640


Add to cart

Description

Description

Connexin 46/GJA3 Polyclonal antibody specifically detects Connexin 46/GJA3 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

Connexin 46/GJA3
Unconjugated
PBS buffer, 2% sucrose
CAE3, connexin 46, connexin-46, Cx46, CX46gap junction alpha-3 protein, CZP3, gap junction protein, alpha 3, 46kD (connexin 46), gap junction protein, alpha 3, 46kDa, gap junction protein, alpha 3, 46kDa (connexin 46)
The immunogen is a synthetic peptide directed towards the C terminal region of mouse Connexin 46/GJA3 (NP_001258552.1). Peptide sequence LQESALVVTPEEGEQALATTVEMHSPPLVLLDPGRSSKSSNGRARPGDLA
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
2700
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit