Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 50/GJA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | Connexin 50/GJA8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1592020
|
Novus Biologicals
NBP15920120UL |
20 μL |
Each for $204.00
|
|
|||||
NBP159201
|
Novus Biologicals
NBP159201 |
100 μL |
Each for $482.50
|
|
|||||
Description
Connexin 50/GJA8 Polyclonal specifically detects Connexin 50/GJA8 in Human samples. It is validated for Western Blot.Specifications
Connexin 50/GJA8 | |
Polyclonal | |
Rabbit | |
P48165 | |
2703 | |
Synthetic peptides corresponding to GJA8(gap junction protein, alpha 8, 50kDa) The peptide sequence was selected from the middle region of GJA8. Peptide sequence EEQEKVAVPEGEKVETPGVDKEGEKEEPQSEKVSKQGLPAEKTPSLCPEL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
alpha 8, 50kD (connexin 50), CAE, Connexin-50, Cx50, CZP1, gap junction alpha-8 protein, gap junction membrane channel protein alpha-8, gap junction protein, alpha 8, 50kDa, gap junction protein, alpha 8, 50kDa (connexin 50), lens intrinsic membrane protein MP70, MP70 | |
GJA8 | |
IgG |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title