Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Contactin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551900
Description
Immunogen sequence: ISNLSVTDSG MFQCIAENKH GLVYSSAELK VVASAPDFSK NPMKKLVQVQ VGSLVSLDCK PRASPRALSS WKKGDVSVQE HERISLLNDG GLKIANVTKA DAGTYTCMAE NQFGKANGTT HLVVTEPTRI Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
Contactin 3 is a type of contactin that mediate cell surface interactions during nervous system development. Has some neurite outgrowth-promoting activity.
Specifications
Contactin 3 | |
Polyclonal | |
Unconjugated | |
CNTN3 | |
BIG-1; Brain-derived immunoglobulin superfamily protein 1; Cntn3; contactin 3; contactin 3 (plasmacytoma associated); contactin-3; KIAA1496; Pang; PCS; Plasmacytoma-associated neuronal glycoprotein; Plasmacytoma-associated neuronal glycoprotein (neural cell adhesion molecule BIG-1) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5067 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P232 | |
CNTN3 | |
Recombinant protein corresponding to Human Contactin 3. Recombinant protein control fragment (Product #RP-102168). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction