Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contactin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific OSC00013W
Description
Glycerol (1:1) may be added for additional stability. Reconstitute with 100 µL of distilled water.
Specifications
Contactin 3 | |
Polyclonal | |
CNTN3 | |
Brain-derived immunoglobulin superfamily protein 1, BIG-1, Plasmacytoma-associated neuronal glycoprotein, KIAA1496, PANG, contactin 3, PCS | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9P232, Q07409, Q62682 | |
CNTN3 | |
A synthetic peptide from c-terminal region of human Contactin 3 conjugated to an immunogenic carrier protein was used as the antigen | |
RUO | |
18488, 5067, 54279 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title