Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Contactin 5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531851
Description
Recommended positive controls: NT2D1, mouse brain, rat brain. Predicted reactivity: Mouse (97%), Rat (95%), Chicken (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
Contactin 5 | |
Polyclonal | |
Unconjugated | |
CNTN5 | |
6720426O10Rik; A830025P08Rik; CNTN5; contactin 5; contactin-5; Gm507; hNB-2; HNB-2s; MGC163491; Nb-2; neural adhesion molecule; neural recognition molecule NB-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114589, 244682, 53942 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.87 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O94779, P68500, P97527 | |
CNTN5 | |
Recombinant fragment corresponding to a region within amino acids 703 and 920 of Human CNTN5. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction