Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Contactin 6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553464
Description
Immunogen sequence: TQEPHDVIFP LDLSKSEVIL NCAANGYPSP HYRWKQNGTD IDFTMSYHYR LDGGSLAINS PHTDQDIGMY QCLATNLLGT ILSRKAKLQF Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development.
Specifications
Contactin 6 | |
Polyclonal | |
Unconjugated | |
CNTN6 | |
CNTN6; contactin 6; contactin-6; hNB-3; mNB-3; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27255 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UQ52 | |
CNTN6 | |
Recombinant protein corresponding to Human Contactin 6. Recombinant protein control fragment (Product #RP-92417). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction