Learn More
Sklar InstrumentsSupplier Diversity Partner SklarLite Sterilization Full Size Container Basket, Sklar, 20 1/2 in. x 10 in. x 3 in., Anodized Aluminum Alloy. SklarLite Sterilization Container Baskets are made from anodized aluminum that allow them to be durable but light. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 104288
SklarLite Sterilization Full Size Container Basket, Sklar, 20 1/2 in. x 10 in. x 3 in., Anodized Aluminum Alloy. SklarLite Sterilization Container Baskets are made from anodized aluminum that allow them to be durable but light. Sklar offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a full size wire basket designed to insert clamps to stabilize instruments during sterilization.
Catalog No. 50-143-7189