Contec™ ABRATEC™ Diamond Grit Abrasives with Quiltec™ Backing
Remediate stainless steel, remove disinfectant residue and minimize rouging due to residues and soils. The Contec™ ABRATEC™ Diamond Grit Abrasives with Quiltec™; Backing are designed to work with non-ketone based solvents. Compatible with autoclave and gamma irradiation processing to help maintain the integrity of sterile environments.
Manufacturer: Contec™ ATQT1350
 View more versions of this product
Catalog No. 19-033-412
  Check Availability
Add to Cart
    Specifications
    Grit Rating 1350
    Material Quiltec™
    Size 8 x 10cm (3 x 4 in.)
    For Use With (Application) Removal of residues from stainless steel surfaces
    •  ABRATEC abrasive cloths are made with various levels of abrasive grits backed with Contec’s unique Quiltec fabric
    • Quiltec fabric is significantly more durable and abrasion resistant than similar foam-backed abrasives
    • The abrasion resistance and the Contec sealed border technology allow the Quiltec side to remove sanding debris without contributing additional particles to the remediation process
    • ABRATEC abrasive cloths are designed to work with solvents, including the IPA/water blends typically found in sterile processing environments
    • ABRATEC cloths are also available irradiated with a dose of 25-50 kGy, which helps maintain the integrity of sterile environments
    • ABRATEC cloths are faster, cleaner, have lower particulation, and are safer than similar foam-backed abrasive products
  • Description & Specifications
Related Products
Product Content Feedback