Learn More
Contec™ ABRATEC™ Diamond Grit Abrasives with Quiltec™ Backing
Remediate stainless steel, remove disinfectant residue and minimize rouging due to residues and soils. The Contec™ ABRATEC™ Diamond Grit Abrasives with Quiltec™; Backing are designed to work with non-ketone based solvents. Compatible with autoclave and gamma irradiation processing to help maintain the integrity of sterile environments.
Supplier: Contec™ ATQT0360
Description
- ABRATEC abrasive cloths are made with various levels of abrasive grits backed with Contec’s unique Quiltec fabric
- Quiltec fabric is significantly more durable and abrasion resistant than similar foam-backed abrasives
- The abrasion resistance and the Contec sealed border technology allow the Quiltec side to remove sanding debris without contributing additional particles to the remediation process
- ABRATEC abrasive cloths are designed to work with solvents, including the IPA/water blends typically found in sterile processing environments
- ABRATEC cloths are also available irradiated with a dose of 25-50 kGy, which helps maintain the integrity of sterile environments
- ABRATEC cloths are faster, cleaner, have lower particulation, and are safer than similar foam-backed abrasive products
Specifications
Removal of residues from stainless steel surfaces | |
Quiltec™ |