Learn More
Contec™ Accel TB™ Ready-to-Use Disinfectants
Ready-to-use one-step disinfectant cleaner
Manufacturer:  Contec™ ACCDISRTB5G
 View more versions of this product
Catalog No. 19-130-5977
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Environmental surfaces in pharmaceutical cleanrooms, compounding pharmacies, labs and other GMP facilities
2.5 to 3.5
18.9L
EPA
Accel TB™ Ready-to-use disinfectant
5 gal.
5 gal., 18.9L
0.5% Hydrogen Peroxide, 99.5% Other ingredients
Safety and Handling
ShelfLife : 2 Year(s)