Contec™ Alcohol-free Foam Hand Sanitizer

Kills up to 99%* of most common germs after a 15 second wet time. Contec™ Alcohol-free Foam Hand Sanitizer is a waterless hand sanitizer and is a better alternative to alcohol-based products. It contains no triclosan or alcohol and leaves hands feeling moisturized and conditioned. Intended for use with Contec™ Alcohol-Free Foam Hand Sanitizer Dispensers.

Manufacturer: Contec™ HAND0200

 View more versions of this product

Catalog No. 19-138-901

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Contec HAND0300, HAND0301, HAND0304, HAND0311
    Capacity 1250mL (42.3 fl. oz.)
    Quantity 6 Bottles/Case
    Product Type Alcohol-free Foam Hand Sanitizer
    Type Benzalkonium chloride formula
    Composition Benzalkonium Chloride
    For Use With (Application) Kills 99.9999% of germs on hands in 15 sec.

    • Contec Alcohol-Free Foam Hand Sanitizer is a waterless hand sanitizer that kills up to 99.9999%* of most common germs after a 15 second wet time
    • A better alternative to alcohol-based products, Contec Alcohol-Free Foam Hand Sanitizer contains no triclosan or alcohol and leaves hands feeling moisturized and conditioned
    • Because it does not contain alcohol, Contec Alcohol-Free Foam Hand Sanitizer will not sting or irritate when applied to skin; this alcohol-free formula also eliminates problems with absorption and/or ingestion
    • CDC guideline compliant E3 classification
    • Contec Alcohol-Free Foam Hand Sanitizer Dispensers sold separately

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit