Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Anticon™ Presaturated Wipes

Avoid the hassle of saturating dry wipes. The Contec™ Anticon™ Presaturated Wipes come with the ideal amount of solvent. Perfect for ISO Class 1 to 4 cleanrooms.

$22.00

Specifications

Dimensions (L x W) 9 x 9 in. (23 x 23 cm)
Material Polyester
View More Specs

Products 1
Catalog Number Mfr. No. Certifications/Compliance Saturation Solution Price Quantity & Availability  
Catalog Number Mfr. No. Certifications/Compliance Saturation Solution Price Quantity & Availability  
19-130-4500
Accessories View Documents
Contec™
495357660
ISO Class 1 to 4 6% Isopropyl Alcohol, 94% Deionized Water
Pack of 50 for $22.00
Case of 16 PK for $352.00
Add to cart
 
Description

Description

  • For ISO Class 1 to 4 (Class 1 to 10) applications
  • Anticon™ Presaturated Wipes are manufactured using ultra-clean Anticon™ Gold and other Anticon™ wipes in conjunction with semiconductor or USP grade solvents
  • These polyester knit cleanroom wipes are presaturated with the optimum amount of solution to deliver absolute cleaning power, not excess solution
  • Anticon™ presats are delivered with a patented low particle generation technology
  • Use Anticon™ presats with the knowledge and security that there are no undo particle burdens being added to the critical environment
  • Made of 100% continuous filament polyester chemically modified for sorbency
  • Premoistened with semiconductor-grade isopropyl alcohol and deionized water; available in five concentrations
  • Sealed edge for low particle generation, less contamination

Compliance

Lot tested in accordance with IEST industry standard

Specifications

Specifications

9 x 9 in. (23 x 23 cm)
Polyester
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit