Learn More
Contec™ Anticon™ Presaturated Wipes
Avoid the hassle of saturating dry wipes. The Contec™ Anticon™ Presaturated Wipes come with the ideal amount of solvent. Perfect for ISO Class 1 to 4 cleanrooms.
Manufacturer:  Contec™ 495357070
 View more versions of this product
Catalog No. 19-150-0293
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • For ISO Class 1 to 4 (Class 1 to 10) applications
  • Anticon™ Presaturated Wipes are manufactured using ultra-clean Anticon™ Gold and other Anticon™ wipes in conjunction with semiconductor or USP grade solvents
  • These polyester knit cleanroom wipes are presaturated with the optimum amount of solution to deliver absolute cleaning power, not excess solution
  • Anticon™ presats are delivered with a patented low particle generation technology
  • Use Anticon™ presats with the knowledge and security that there are no undo particle burdens being added to the critical environment
  • Made of 100% continuous filament polyester chemically modified for sorbency
  • Premoistened with semiconductor-grade isopropyl alcohol and deionized water; available in five concentrations
  • Sealed edge for low particle generation, less contamination
Description & Specifications
Specifications
Suitable for tool chamber, wall wipe downs, and pre-gown wipe down
23cm
ISO Class 1 to 4
9 in.
70% Isopropyl Alcohol, 30% Deionized Water
9 in.
9 in.
Anticon™ presaturated wipes
9 x 9 in. (23 x 23cm)
Polyester
9 in.
23cm
Accessories