Learn More
Contec™ Bucket Wringers and Frames
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Manufacturer:  Contec™ 2765
 View more versions of this product
Catalog No. 19-033-716
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
  • Choose from downpress, flat or curved styles
  • Available in polypropylene or stainless steel
  • Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
  • Select wringer that complements mop
Specifications
Specifications
Contec™ flat or curved bucket wringers; for 5 gal. (20L) buckets only
Bucket wringer frame
Stainless steel