Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ CONSTIX™ Cotton Swabs

Manufactured with the longest staple cotton fibers and a simple methyl-cellulose binder. Contec™ CONSTIX™ Cotton Swabs provide very low levels of extractable residue, particles and fibers.

Manufacturer:  Contec™ SC2

 View more versions of this product

Catalog No. 19-130-1519


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.1 in.
2.5mm
For use in some of the most critical applications, including disk drive manufacturing and high-level optical surfaces
3 in.
76.2mm
Cotton
0.17 in.
4.3mm
Swab
0.2 in.
5.1mm
Wound Paper
0.60 in.
15.2mm
Round double ended
0.17 in.
4.3mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit