Learn More
Contec™ EasyCurve™ Mop Heads
Choose from non-sterile or validated sterile versions; mop heads are autoclavable. The Contec™ EasyCurve™ Mop Heads use curved mop faces with pivoting connectors for easier surface contact at a variety of angles. 
Manufacturer:  Contec™ EZMP0100
 View more versions of this product
Catalog No. 19-168-746
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
EasyCurve™ Mopping System provides the convenience of a self-wringing mop without the ergonomic pain. Compact system (mop, bucket and cart) is designed for all size cleanrooms and requires minimal storage space. Mop heads attach to a curved, stainless steel frame  to provide superior performance, flexibility, and reach. The curved head design offers a true "lift and pull" surface cleaning action, ensuring uniform distribution of disinfectants and cleaners. All components are sold separately.
  • Flat, fabric-laminated mop heads 
  • Drip-free hydrophillic polyurethane foam; compatible with most common solvents
  • Durable polyester fabric outer layer reduces particulation and ensures uniform distribution of disinfectants and cleaners
  • Non-dripping mop head suitable for use on ceilings, walls and floors
  • Sterile or nonsterile
  • Available with a sealed edge to increase durability and eliminate the chance of fiber and particle contamination due to exposed foam
  • Ribbed microfiber face provides users with smooth and easy mopping for better particle pick-up and contamination removal
Specifications
Specifications
Non-sterile
Yes
Mop head, polyester
Designed for use in pharmaceutical and medical device manufacturing plants, hospitals, compounding pharmacies and other critical or controlled environments
14 x 4.25 x 0.75 in. (36 x 11 x 2cm)
4/bag, 12 bags/case
Knitted polyester fabric / hydrophilic urethane foam
Yes
EasyCurve mop system