Contec™ EasyCurve™ Mop Heads
Choose from non-sterile or validated sterile versions; mop heads are autoclavable. The Contec™ EasyCurve™ Mop Heads use curved mop faces with pivoting connectors for easier surface contact at a variety of angles. 
Manufacturer: Contec™ EZMP0150
 View more versions of this product
Catalog No. 19-168-747
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Mop head, polyester
    Disposable Yes
    Quantity 1/bag, 48 bags/case
    Material Knitted polyester fabric / hydrophilic urethane foam
    For Use With (Application) Designed for use in pharmaceutical and medical device manufacturing plants, hospitals, compounding pharmacies and other critical or controlled environments
    For Use With (Equipment) EasyCurve mop system
    Size 14 x 4.25 x 0.75 in. (36 x 11 x 2cm)
    Autoclavable Yes
    Sterile Yes
    EasyCurve™ Mopping System provides the convenience of a self-wringing mop without the ergonomic pain. Compact system (mop, bucket and cart) is designed for all size cleanrooms and requires minimal storage space. Mop heads attach to a curved, stainless steel frame  to provide superior performance, flexibility, and reach. The curved head design offers a true "lift and pull" surface cleaning action, ensuring uniform distribution of disinfectants and cleaners. All components are sold separately.
    • Flat, fabric-laminated mop heads 
    • Drip-free hydrophillic polyurethane foam; compatible with most common solvents
    • Durable polyester fabric outer layer reduces particulation and ensures uniform distribution of disinfectants and cleaners
    • Non-dripping mop head suitable for use on ceilings, walls and floors
    • Sterile or nonsterile
    • Available with a sealed edge to increase durability and eliminate the chance of fiber and particle contamination due to exposed foam
    • Ribbed microfiber face provides users with smooth and easy mopping for better particle pick-up and contamination removal
  • Description & Specifications
Related Products
Product Content Feedback