Contec™ EasyReach™ Cleaning System

Passes easily through transfer hatches, requires no assembly and is maintenance-free. Contec™ EasyReach™ Cleaning System is ideal for isolators, cabinets, laminar air-flow hoods, glove boxes and other mini environments.

Manufacturer: Contec™ MEQT0002

 View more versions of this product

Catalog No. 19-830-002

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) • Isolators - rigid, flexible film, half suits (CAI, CACI) • Restricted Access Barrier Systems (RABS) • Filling Lines • Laminar Air Flow Hoods (LAF) • Bio-Safety Cabinets (BSC) • Glove Boxes • Incubators
    Material Quilted polyester
    Product Type Cleaning pad
    Disposable Yes
    Length (English) 7.5 in.
    Width (English) 2.7 in.
    Autoclavable No
    Sterile Yes

    EasyReach™ Cleaning System consists of a lightweight stainless steel 1 piece cleaning tool and interchangeable disposable and absorbant cleaning pads that provide effective cleaning and disinfection in hard to reach areas. Components are sold separately

    • Rectangular shape makes overlapping straight-line action easy to achieve
    • Low profile 360° pivoting head allows quick easy access to awkward areas
    • Hinged joint and lightweight design
    • Easy grab edge on the cleaning pad ensures pad fits tightly on the mop head frame; easy to replace with gloved hands
    • Small pack quantities are optimized for single session use
    • Well-fitted quilted polyester pad ensures effective, even surface coverage, reducing the number of viable and non-viable contaminants and ensuring maximum residue removal
    • Sorbent double layer Quiltec™ sealed-edge pads ensure particles and fibers are minimized
    • Cleaning pads available presaturated with 70% IPA 
    See QuickConnect™ Mop head frames and handles for long handled EasyReach option.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit