Contec™ Edgeless™ Mopping Systems
Available as standard mop, or with enhanced ribbed tube mop head
Manufacturer: Contec™ ELTCBLU
 View more versions of this product
Catalog No. 19-033-852
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
    • Mop head is a continuous tube of 100% polyester-knitted fabric looped to form tubular mop strands—compatible with a wide range of chemicals
    • Mop strands encapsulate liquids for maximum sorption; generates significantly lower particulate and extractable levels compared to conventional cotton string or nylon mops
    • Mop head also available sterilized by gamma radiation; can also be sterilized by EtO and steam (autoclave) sterilization methods
    • Validated sterile per the AAMI/ISO Guidelines to a 10-6 Sterility Assurance Level (certificates of sterility available)
    • Mop heads laundered and packaged in an ISO Class 4 cleanroom
    • Autoclavable plastic double bucket system features a wringer and cart with casters
    • Mop head handle, Dynamate™ frame (19-130-5911) and bucket system sold separately
  • Description & Specifications
Related Products
Product Content Feedback