Contec™ Edgeless™ Mopping Systems
Available as standard mop, or with enhanced ribbed tube mop head
Manufacturer: Contec™ ELTCGRN
 View more versions of this product
Catalog No. 19-033-855
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Application) Aqueous solutions containing cleaning agents and disinfectants
    Autoclavable Yes
    Material Textured; Green tail band: Knitted Polyester
    Product Type Mopping System
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback