Contec™ Edgeless™ Mop System
Clean in controlled environments and cleanrooms. The Contec™ Edgeless™ Mop System is constructed from a continuous tube of 100% polyester knitted fabric. The mop heads help eliminate contamination from breakage and deterioration of traditional string mops.
Manufacturer: Contec™ 2695
Catalog No. 19-168-745
  Check Availability
Add to Cart
    Specifications
    Sterile No
    For Use With (Equipment) Edgeless mop system
    Autoclavable Yes
    Product Type Wringer Insert
    Material Polypropylene
    • Designed for wet mopping in controlled environments and cleanrooms
    • Ideal for use in sterile environments
    • Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption
    • Mop heads are free of any cellulosic contaminants
    • Textured mop heads feature ribbed, twisted strands for 60% more sorbent capacity than standard Edgeless mops
    • Textured knitted polyester tube mop heads help eliminate contamination from breakage and deterioration of traditional string mops
    • Mop heads are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom
  • Description & Specifications
Related Products
Product Content Feedback