Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Polynit Heatseal Wipes

Laundered and packaged in an ISO Class 4 cleanroom. Contec™ Polynit Heatseal Wipes are available with Particle Attraction Technology (PAT).

Manufacturer:  Contec™ LWPS0052

 View more versions of this product

Catalog No. 19-170-933

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

 

  • 100% textured polyester wipe with nonabrasive, heat-sealed edge
  • Chemical- and abrasion-resistant, with good solvent sorbency
  • Extremely low in both particles, fibers, and nonvolatile residues
  • Specially treated to enhance particle attraction and retention
  • Attracts 35 times more particles than traditional wipes
  • Retains 95% of particles captured, so there is less chance of re-contamination

 

Wipes with Particle Attraction Technology (PAT)

  • 19-170-930: 9 x 9 in.
  • 19-170-931: 12 x 12 in.
  • 19-170-932: 4 x 4 in.
  • 19-170-933: 9 x 9 in.

 

Applications:

Ideal for general-purpose cleaning and product wipe down.

Specifications

Specifications

ISO Class 4
Ideal for general purpose cleaning and product wipe down
23 cm
Polynit heatseal wipes with P.A.T
150 wipes/bag, Bulk
9 in.
9 x 9 in. (23 x 23 cm)
9 in.
Polyester
9 in.
9 in.
23 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit