Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Polynit™ Heatseal Wipes Get IT Item Available on GSA/VA Contract for Federal Government customers only.

Clean the exterior of products and cleanroom surfaces. Contec™ Polynit™ Heatseal Wipes are made of 100% textured polyester with laser-cut sealed edges to prevent dust and residue. They are extremely low in particles, fibers and non-volatile residues (NVRS).

Manufacturer:  Contec™ PNHS99B/150

 View more versions of this product

Catalog No. 19-047-513



  Check Availability
Add to cart

Specifications

Description

  • Textured 100% polyester no-run knit wipes with heatsealed edges
  • Laser-cutting seals fibers at wipe's edge and allows for 100% utilization of wipe
  • Laundered and packaged in a ISO Class 4 cleanroom
  • Extremely low in particles, fibers and non-volatile residues (NVRS)
  • Good sorbency with solvents
  • Abrasion and chemical resistant
Description & Specifications

Specifications

140
9 x 9 in. (23 x 23cm)
Polyester
No
9 in.
Ideal for general purpose cleaning
23cm
ISO Class 4
9 in.
150 wipes/bag, Bulk
9 in.
23cm
9 in.
Polynit Heatseal wipes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit