Learn More
Contec™ SATWipes™ Presaturated Rayon Wipes
Prepare for a variety of cleanrooms. The Contec™ SATWipes™ Presaturated Rayon Wipes are offered in various options to help tackle general-purpose cleaning in ISO Class 5 to ISO Class 8 environments.
Manufacturer:  Contec™ SW420902
 View more versions of this product
Catalog No. 19-159-232
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Durable, presaturated wipers with a choice of solvent saturations on perforated rolls
  • SATWipes with Polyester Nonwoven wipers are ideal for critical applications in ISO Class 5 environments
  • SATWipes with Polyester/Cellulose Nonwoven Wipers offer strength and low extractables for use in less critical applications in ISO Class 6 to 8 environments
  • Designed for general-purpose cleaning and surface preparation and for wiping process tools and other equipment and for use prior to cleanroom entry
Description & Specifications
Specifications
9 x 6 in. (23 x 15.2cm)
Methyl Propyl Ketone
15.2cm
23cm
Rayon
6 in.
9 in.
Rayon Satwipes™ presaturated wipes