Learn More
Contec™ SATWipes™ Presaturated Wipes
Prepare for a variety of cleanrooms. The Contec™ SATWipes™ Presaturated Wipes are offered in various options to help tackle general-purpose cleaning in ISO Class 5 to ISO Class 8 environments.
Manufacturer:  Contec™ SWC30081
 View more versions of this product
Catalog No. 19-170-951
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Durable, presaturated wipes with a choice of solvent saturations on perforated rolls
  • SATWipes with Polyester Nonwoven wipes are ideal for critical applications in ISO Class 5 environments
  • SATWipes with Polyester/Cellulose Nonwoven Wipes offer strength and low extractables for use in less critical applications in ISO Class 6 to 8 environments
  • Designed for general-purpose cleaning and surface preparation and for wiping process tools and other equipment and for use prior to cleanroom entry
Description & Specifications
Specifications
ISO Class 5 and 6
Polyester
70% Isopropyl Alcohol, 30% Deionised Water
10.16cm
4.25 in.
SATWipes presaturated wipes
4.25 x 4 in. (10.7 x 10.16cm)
185 wipes/roll
4 in.
Excellent wipes for less critical applications, compatible with ISO class 6 environments and wipes down prior to entry into cleanroom
10.7cm