Contec™ Sporicidin™ Sterilizing and High Level Disinfectant
Sporicidin Sterilizing and High Level Disinfecting Solution is intended to sterilize or high level disinfect reusable medical instruments and equipment
Manufacturer: Contec™ PMG-6000
 View more versions of this product
Catalog No. 18-999-1458
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Indicator kit
    • Sporicidin Sterilizing and High Level Disinfecting Solution is intended to sterilize or high level disinfect reusable medical instruments and equipment 
    • Requires no rotation
    • It also has the shortest soaking time (20 minutes) for high level disinfection
    • Can be used and reused for 14 days
    • Due to low glutaraldehyde level, will not irritate the eyes or nasal passages
    • Will not clog air/water channels or cloud lenses of endoscopes
    • OSHA compliant with Bloodborne Pathogens Standard
    • Environmentally safe
    • Dispose in regular drain
  • Description & Specifications
Related Products
Product Content Feedback