Learn More
Contec™ Sporicidin™ Sterilizing and High Level Disinfectant
Sporicidin Sterilizing and High Level Disinfecting Solution is intended to sterilize or high level disinfect reusable medical instruments and equipment
Manufacturer:  Contec™ SSS1284C
 View more versions of this product
Catalog No. 18-999-1456
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Sporicidin Sterilizing and High Level Disinfecting Solution is intended to sterilize or high level disinfect reusable medical instruments and equipment 
  • Requires no rotation
  • It also has the shortest soaking time (20 minutes) for high level disinfection
  • Can be used and reused for 14 days
  • Due to low glutaraldehyde level, will not irritate the eyes or nasal passages
  • Will not clog air/water channels or cloud lenses of endoscopes
  • OSHA compliant with Bloodborne Pathogens Standard
  • Environmentally safe
  • Dispose in regular drain
Description & Specifications
Specifications
Sporicidin™ sterilizing and high level disinfecting solution is intended to sterilize or high level disinfect reusable medical instruments and equipment
1 gal.
1 gal., 3.8L
1.93% Phenol, 1.12% Glutaraldehyde
Sterilizing and disinfecting solution
3.8L
Clear
Sporicidin™
Safety and Handling
ShelfLife : 14 Day(s)