Learn More
Contec™ Quiltec™ I Presaturated Wipes, sterile
BioPharma
Provide a stronger, more durable wipe that is chemical and abrasion resistant. Quiltec™ I Presaturated Wipes, sterile are saturated with a solution of isopropyl alcohol and deionized water. They are extremely low in particle and fiber generation.
Specifications
Sterility Sterile
View More Specs
Products 1
Catalog Number Mfr. No. Dimensions (L x W) Material Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Material Price Quantity & Availability  
19-150-0708 BioPharma
Accessories View Documents
Contec™
PSQS0001
12 x 12 in. Quiltec Polyester
Case of 300 for $537.17
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Knitted polyester wipes presaturated with 70% USP grade IPA/30% DI water
  • Validated sterile at a 10-6 SAL per AAMI/ISO 11137-2:2012 Guidelines
  • Ideal for wiping of surfaces and equipment
  • Extremely low in particle and fiber generation
  • Resealable packages
Order Info
ISO Class 4 Cleanroom laundered and packaged in resealable packs. 19-120-816 packed 20 wipers/pack; 19-160-965 and 19-150-0708 are half folded in packs of 30; sold in case quantities only.
Specifications
Specifications
Sterile