Contec™ Tax-Fre™ Dry Mopping System
Dry mop floors and walls in cleanroom, laboratory, clinical and industrial settings. The Tax-Fre™ Dry Mopping System, Contec™ features a telescoping handle for hard to reach places. Its pressure-sensitive formulation will not scratch or damage surfaces.
Manufacturer: Contec™ WWP08
 View more versions of this product
Catalog No. 19-150-0703
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Ideal for cleaning floors and walls in powder spray environments
    Length (English) 8 ft.
    Material Aluminum
    Product Type Extension Mop Handle
    • Disposable covers–with unique, non-transfering, low-tack adhesive–capture small particles (powder, dander, dust, lint, hair and other debris)
    • Extremely clean surfaces without the use of solvents or other liquids
    • Covers feature soft texture;  will not scratch or harm sensitive surfaces
    • Large trapezoid-shaped mop head swivels on a lightweight 2.4m (8’) telescoping aluminum handle, allowing operators to clean hard-to-reach areas
    • Designed for dry mopping floors and walls when liquids are not recommended
    • Large 23 x 5 in. (58.4 x 13cm) tapered swivel head attaches to lightweight 8 ft. (2.4m) telescoping handle (two-piece) allowing operator to clean hard-to-reach areas
    • Pressure-sensitive formulation will not scratch or damage surfaces
  • Description & Specifications
Related Products
Product Content Feedback