Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Contec™ Edgeless™ Non-Sterile Mop Heads
Manufactured from a continuous tube of strong, durable 100% polyester knitted fabric. Contec™ Edgeless™ Non-Sterile Mop Heads are looped to form tubular mop strands that will not shred, break or deteriorate, eliminating contamination.
Manufacturer:  Contec™ EL5GRN
 View more versions of this product
Catalog No. 19-033-963
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
 The Contec™ Edgeless™ Mopping System is the cleanest floor mopping system available for critical environments. The system is designed for semiconductor, pharmaceutical, biotech and medical device industries for lab clean up and process area cleaning. Mop heads are designed to meet general purpose cleaning needs in ISO Class 3 or less critical environments and are available in a wide variety of models to meet different cleaning needs. Edgeless™ or QuickConnect™ handles, frames, adaptors and bucket systems are available separately to complete the system.
  • Mop strands encapsulate liquids for maximum sorption; generate significantly lower particulate and extractable levels compared to conventional cotton string or nylon mops
  • Mop heads laundered and packaged in an ISO Class 4 cleanroom
  • Resistant to chemicals, mold and mildew
  • Loops eliminate cut edges and provide improved cleanliness
  • Tail bands improved control and allow easy mopping around equipment legs and wheels
  • Available in four models:
    • Original--traditional tub construction
    • Textured--twisted strands increase sorbent capacity
    • Long textured--holds more liquid for mopping larger areas; lasts up to 10x longer than traditional mops; feature colored tail bands for area segregation to minimize cross-contamination
    • Enhanced ribbed tube mop head with textured headband--increased durability
Description & Specifications
Specifications
White with green tail band
Knitted Polyester
No
Contec Edgeless Mopping System
Yes
Non-sterile cleanroom laundered long textured mop head
Designed for use with aqueous solutions containing cleaning agents and disinfectants; compatible with wide range of chemicals