Learn More
Contec™ Alcohol-free Foam Hand Sanitizer Dispensers
BIOPHARMA
Dispense Contec™ Alcohol-Free Foam Hand Sanitizer, which kills up to 99%* of most common germs after a 15 second wet time. Contec™ Alcohol-free Foam Hand Sanitizer Dispensers are available in manual or touch-free options. They are also available in a variety of colors.
Manufacturer:  Contec™ HAND0301
 View more versions of this product
Catalog No. 19-138-904
Add to cart
This item is not returnable. View return policy
Description
Description
  • Dispensers available in touch-free or manual varieties
  • Optional drip tray available
  • Contec Alcohol-Free Foam Hand Sanitizer sold separately
Contec Alcohol-Free Foam Hand Sanitizer
  • Waterless hand sanitizer that kills up to 99%* of most common germs after a 15 second wet time
  • A better alternative to alcohol-based products, Contec Alcohol-Free Foam Hand Sanitizer contains no triclosan or alcohol and leaves hands feeling moisturized and conditioned
  • Because it does not contain alcohol, it will not sting or irritate when applied to skin; this alcohol-free formula also eliminates problems with absorption and/or ingestion
  • Each cartridge has approximately 1650 applications
  • CDC guideline compliant E3 classification
Specifications
Specifications
CDC guideline compliant E3 classification
Touchless Dispenser
Hand sanitizer cartridges: Contec HAND0200, HAND0201
White
Non-flammable and non-hazardous for easy transport/storage
One set of batteries
SDS
Documents
Product Certifications
Accessories