Learn More
Contec™ Alcohol-free Foam Hand Sanitizer Dispensers
Dispense Contec™ Alcohol-Free Foam Hand Sanitizer, which kills up to 99%* of most common germs after a 15 second wet time. Contec™ Alcohol-free Foam Hand Sanitizer Dispensers are available in manual or touch-free options. They are also available in a variety of colors.
Supplier: Contec™ HAND0304
Description
- Dispensers available in touch-free or manual varieties
- Optional drip tray available
- Contec Alcohol-Free Foam Hand Sanitizer sold separately
- Waterless hand sanitizer that kills up to 99%* of most common germs after a 15 second wet time
- A better alternative to alcohol-based products, Contec Alcohol-Free Foam Hand Sanitizer contains no triclosan or alcohol and leaves hands feeling moisturized and conditioned
- Because it does not contain alcohol, it will not sting or irritate when applied to skin; this alcohol-free formula also eliminates problems with absorption and/or ingestion
- Each cartridge has approximately 1650 applications
- CDC guideline compliant E3 classification
Compliance
NSF approved E3 classification for use around food processing areas
Specifications
White | |
Non-flammable and non-hazardous for easy transport/storage | |
1 Ea. |
NSF approved E3 classification | |
Hand sanitizer cartridges: Contec HAND0200, HAND0201 |