Contec™ Alcohol-free Foam Hand Sanitizer Dispensers

Dispense Contec™ Alcohol-Free Foam Hand Sanitizer, which kills up to 99%* of most common germs after a 15 second wet time. Contec™ Alcohol-free Foam Hand Sanitizer Dispensers are available in manual or touch-free options. They are also available in a variety of colors.

Manufacturer: Contec™ HAND0300

 View more versions of this product

Catalog No. 19-138-903

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Hand sanitizer cartridges: Contec HAND0200, HAND0201
    Certifications/Compliance CDC guideline compliant E3 classification
    Color Black
    For Use With (Application) Non-flammable and non-hazardous for easy transport/storage
    Includes One set of batteries
    Product Type Touchless Dispenser

    • Dispensers available in touch-free or manual varieties
    • Optional drip tray available
    • Contec Alcohol-Free Foam Hand Sanitizer sold separately
    Contec Alcohol-Free Foam Hand Sanitizer
    • Waterless hand sanitizer that kills up to 99%* of most common germs after a 15 second wet time
    • A better alternative to alcohol-based products, Contec Alcohol-Free Foam Hand Sanitizer contains no triclosan or alcohol and leaves hands feeling moisturized and conditioned
    • Because it does not contain alcohol, it will not sting or irritate when applied to skin; this alcohol-free formula also eliminates problems with absorption and/or ingestion
    • Each cartridge has approximately 1650 applications
    • CDC guideline compliant E3 classification

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit