Learn More
Contec™ VertiKlean™ MAX™ Mopping Systems Mop Heads
Clean walls, ceilings and other hard-to-reach surfaces with ease. The Contec™ VertiKlean™ MAX™ Mopping Systems Mop Heads feature a variety of heads that easily snap onto frames. The polyurethane foam lasts longer than traditional mops and more evenly releases liquid.
Manufacturer:  Contec™ VKMX0200
 View more versions of this product
Catalog No. 19-165-65
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Features a single layer of hydrophilic polyurethane foam laminated to textured polyester microfiber fabric
  • Backed by a semi-rigid thermoformed plate that easily snaps onto mop head frame 
  • Designed for applying disinfectants to both horizontal and vertical surfaces
  • Mop heads are composed of a disposable absorbent sponge laminated to filament polyester knit fabric and backed with a thermoformed plate that easily snaps onto VertiKlean mop hardware
  • Now available with ribbed microfiber for better contamination removal
  • Pivoting mop head frame is ideal for cleaning walls, floors, and ceilings as well as hard-to-reach areas such as fume hoods, mini environments, and around light fixtures and heavy equipment
  • Sterile mop heads are validated sterile per AAMI ISO11137 guidelines
  • Autoclaving mop heads is not recommended due to thermoformed plate; however, all hardware is autoclavable
Now available with a sealed edge
  • Thermally sealed edges of the mop head increase durability and eliminate the chance of fiber and particle contamination due to exposed foam
  • Sealed Edge mop head has a microfiber face providing excellent removal of surface contamination and residue
Description & Specifications
Specifications
Medium VertiKlean MAX mop head
Knitted polyester fabric/ 100ppi hydrophilic urethane foam
No
Wall washing system
12.8 in.
12.8 x 4.3 in. (32 x 11cm)
AAMI ISO 11137
Mop Head
Yes
0.7 in.
4.3 in.