Learn More
Contec™ QuickConect™ Mop Head Frames
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Manufacturer:  Contec™ 2676
 View more versions of this product
Catalog No. 19-038-555
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
QuickConnect™ mop head frames and handles offer maximum flexibility for cleaning in critical environments. Invest in one handle and choose various interchangeable heads and frames for specific applications.
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
  • Capped end and quick-connect feature for easy attachment
Description & Specifications
Specifications
Designed for use in pharmaceutical and medical device manufacturing facilities, or other similar critical or controlled environments.
Replacement for regular mop head frame for VertiKlean, VertiKlean MAX
19cm
Anodized aluminum
Regular VertiKlean and VertiKlean MAX mop heads
7.5 in.
Yes
Mop head frame