Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contemporary Chemistry
From essential chemical concepts and ideas to the importance of chemistry in daily life.
$23.00
Specifications
For Use With (Application) | Chemical concepts and ideas to the importance of chemistry in daily life for students |
---|---|
Format | Book |
Category | Chemistry |
Max. Grade Level | 12 |
Min. Grade Level | 9 |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S67874
![]() |
Walch Education![]() 023186 |
Contemporary Chemistry: A Practical Approach, Teachers Guide |
Each for $23.00
|
|
Description
- Provides assessments and helps develop critical-thinking and problem-solving skills
- Lesson plans
- Discussion questions
- Strategies for building reading, writing, and math skills
- Leonard Saland, Paperback, 1991
- Grades 9-12
Specifications
Chemical concepts and ideas to the importance of chemistry in daily life for students | |
Chemistry | |
9 |
Book | |
12 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title