Learn More
Crystal Chem IncSupplier Diversity Partner Control for the Human Total Bile Acid (Enzyme Cycling) Kit DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80253
The Control for the Total Bile Acid Assay (Enzyme Cycling) can be used to validate the calibration curve obtained when running the Human Total Bile Acid Assay (Enzyme Cycling). Use this product when: running the assay for the first time, using the kit in a novel way, or when checking the activity of a stored kit.
Catalog No. 50-194-7679