Learn More
Dynamic Diagnostics Control Freek Working Vial Rack
Control Freek Working Vial Racks are a great help for organizing your control vials with our unique swiveling vial rack.
Manufacturer:  Dynamic Diagnostics 2284
Catalog No. 22-899-669
Add to cart
Description
Description
  • Wedges sit securely on the unique swiveling vial rack base
  • Each rack comes with four pizza slice shaped removable wedges
  • The Control Freek allows you to organize controls and calibrator chemicals in a manageable way – each rack swivels like a Lazy Susan for quick access
  • Each pizza slice is individually colored and can be removed to sit next to a particular analyzer, or can be independently put into a refrigerator, etc
  • Easily stores, segregates and organzies your controls and cailbrators so they are easy to retrieve
  • Each segment has three 30mm and five 24mm holes for vials
  • Made of durable, easy to clean acrylic
Specifications
Specifications
Healthcare
Multicolored
32
Not Autoclavable
Acrylic
1