Learn More
Thermo Scientific™ Convenience Kit, Certified 9mm Wide Opening SureStop Vials with ID Patch PROMO
Use certified clean, low particle, low background vials for your high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
Specifications
Color Clear
Closure Color Blue
Material Glass, PTFE, Silicone
No. per Pack 100
View More Specs
Products 2
Catalog Number Mfr. No. Septum Pre-slit Price Quantity & Availability  
Catalog Number Mfr. No. Septum Pre-slit Price Quantity & Availability  
03-151-692PM
View Documents
Thermo Scientific™
CERT5000580WPR
Ivory PTFE/Red Rubber No This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-052-450PM
View Documents
Thermo Scientific™
CERT5000593WPR
Blue PTFE/White Silicone Yes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Unassembled kits
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers
Assembled kits
  • 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers
Specifications
Specifications
Clear
Blue
Glass, PTFE, Silicone
100